Learn More
Neuromics, Inc Strep-Tactin HRP Conjugate, 0.5 ml is specifically optimized for binding to Strep-Tag II and Twin-Strep-tag fusion proteins.
Supplier:  Neuromics, Inc 21502001
Encompass
Strep-Tactin labeled with peroxidase (2- 2.5 moles per mole Strep-Tactin). Strep-Tactin* is a streptavidin variant which has been specially optimized for the binding to Strep-tagII and Twin-Strep-tag fusion proteins
Catalog No. NC2071878