Learn More
IBA LifeSciences Strep-Tactin Superflow resin as 50% suspension; 50 ml
Supplier:  IBA LifeSciences 21206025
Encompass_Preferred
Strep-Tactin Superflow is a 6% agarose coupled with the streptavidin variant Strep-Tactin and applicable for purification of Strep-tagII or Twin-Strep-tag fusion proteins. The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification. To pack a column with 1 ml resin bed volume, 2 ml 50% suspension is necessary.
Catalog No. NC0105865