Learn More
IBA LifeSciences Strep-TactinXT 4Flow cartridge; 1 ml
Catalog No. 502100154
Encompass_Preferred
Strep-TactinXT 4Flow cartridges contain a 4% agarose coupled with the streptavidin variant Strep-TactinXT and are applicable for purification of Strep-tagII or Twin-Strep-tag fusion proteins with HPLC/FPLC devices like Äkta systems (Cytiva). Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and highest binding affinity, the resin is recommended for all protein classes, especially for low abundant or challenging proteins. The low concentrated agarose enables the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets. In addition, Strep-TactinXT 4Flow allows the application of denaturing agents such as urea.
Catalog No. 50-210-0154 Supplier IBA LifeSciences Supplier No. 25023001
Strep-TactinXT 4Flow cartridges contain a 4% agarose coupled with the streptavidin variant Strep-TactinXT and are applicable for purification of Strep-tagII or Twin-Strep-tag fusion proteins with HPLC/FPLC devices like Äkta systems (Cytiva). Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and highest binding affinity, the resin is recommended for all protein classes, especially for low abundant or challenging proteins. The low concentrated agarose enables the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets. In addition, Strep-TactinXT 4Flow allows the application of denaturing agents such as urea.