Learn More
IBA LifeSciences Strep-TactinXT 4Flow HC
Supplier:  IBA LifeSciences 25030500
Encompass
Strep-TactinXT 4Flow high capacity is perfectly suited for purifying Strep-tagII or Twin-Strep-tag fusion proteins. This resin offers excellent pressure stability, pH compatibility, and target purity (> 95%). Additionally, this resin is compatible with a broad range of reagents.Its high binding affinity makes it ideal for low abundant, high-expressing and challenging proteins. A low concentration of agarose also facilitates the purification of large proteins (>90 kDa). When working with, for example, multimeric proteins, choosing a resin with the Strep-Tactin ligand is recommended.Strep-TactinXT 4Flow high capacity resin has a higher density of immobilized Strep-TactinXT than the classic Strep-TactinXT 4Flowand thus can bind more protein.As the resin is optimized for column affinity chromatography, rather than batch purification, the suspension can be used to pack gravity flow columns.
Catalog No. NC3566336
Add to Cart
Request a Quote