Learn More
Neuromics, Inc Strep-TactinXT 4Flow Resin, 20 ml is used for purification of recombinant Strep-tagII and Twin-Strep-tag fusion proteins.
Catalog No. NC2071883
Encompass
Strep-TactinXT 4Flow is used for purification of recombinant Strep-tagII and Twin-Strep-tag fusion proteins under gravity flow and low pressure or FPLC.Dynamic binding capacity: 7 mg protein/ml resin, determined with mCherry-Twin-Strep-tag (30kDa).
Catalog No. NC2071883 Supplier Neuromics, Inc Supplier No. 25010010
Strep-TactinXT 4Flow is used for purification of recombinant Strep-tagII and Twin-Strep-tag fusion proteins under gravity flow and low pressure or FPLC.Dynamic binding capacity: 7 mg protein/ml resin, determined with mCherry-Twin-Strep-tag (30kDa).