Learn More
Germaine™ Laboratories StrepAim™ Direct Group A Streptococcus Antigen Strip
Catalog No. 23111375
Change view
Click to view available options
Quantity:
25 Tests/Box
Catalog No. Quantity
23-111-375 25 Tests/Box
1 options
This item is not returnable. View return policy
Catalog No. 23-111-375 Supplier Germaine™ Laboratories Supplier No. 73025
Add to Cart
Add to Cart
This item is not returnable. View return policy
A rapid immunochromatographic assay for the qualitative detection of group A streptococcal antigen directly from throat specimens
  • Intended for use in physician offices, hospitals, and clinical laboratories as an aid in the clinical diagnosis
  • Convenient workstation
  • Dipsticks, disposable test tubes, sterile swabs
  • Positive and negative controls
  • CLIA Waved
Specifications
Accuracy 99%
Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Streptococci (Group A)
Disposable Single-use
Quantity 25 Tests/Box
Test Time 5 min.