Learn More
Germaine™ Laboratories StrepAim™ Direct Group A Streptococcus Antigen Strip
Description
- Intended for use in physician offices, hospitals, and clinical laboratories as an aid in the clinical diagnosis
- Convenient workstation
- Dipsticks, disposable test tubes, sterile swabs
- Positive and negative controls
- CLIA Waved
Specifications
Specifications
| Content And Storage | 2°C to 30°C |
| Clia Complexity | Waived |
| Detectable Analytes | Streptococci (Group A) |
| Sample Type | Throat |
| Disposable | Single-use |
| Quantity | 40 Tests/Box |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.