Learn More
IBA LifeSciences StrepMan Magnet
Catalog No. NC3566470
Encompass_Preferred
The StrepMan consists of a plastic frame with two embedded permanent magnets and accommodates two different tube sizes (2x 15 ml and 2x 50 ml conical reaction tubes). It can be used for magnetic cell isolation with microbeads such as Strep-Tactin Magnetic Microbeads.
Catalog No. NC3566470 Supplier IBA LifeSciences Supplier No. 65650065
The StrepMan consists of a plastic frame with two embedded permanent magnets and accommodates two different tube sizes (2x 15 ml and 2x 50 ml conical reaction tubes). It can be used for magnetic cell isolation with microbeads such as Strep-Tactin Magnetic Microbeads.