Learn More
Description
- Accurate results in one minute at room temperature
- Utilizes modified acid extraction technology
- No incubation is required
- All color-coded reagents are ready-to-use and remain in one space-saving workstation
- Easy to read blue latex particles on a white reaction card with eight reaction circles create an exceptionally easy-to-read result
- Greater than 98% sensitive with as little as one colony required
- A complete procedure takes only 2 minutes
- Kit contains reaction cards and mixing sticks
- Provided in a kit format or separate components
Specifications
Specifications
| Detectable Analytes | Streptococci (Group A, B, C, D, F, G) |
| Type | Agglutination |
| Disposable | Single-use |
| No. of Reactions | 60 |
| Analysis Time | 1 min. |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
