Learn More
Hardy Diagnostics™ StrepQuick™ To Aid in the Identification of Gram-positive Cocci
SDP
Catalog No. 23001758 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Analysis Time:
10 min.
Catalog No. Analysis Time
23-001-758 10 min.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-758 Supplier Hardy Diagnostics™ Supplier No. Z122
Add to Cart
Add to Cart
This item is not returnable. View return policy
StrepQuick™ test kit is intended to aid in the identification of gram-positive, catalase-negative cocci based on pyroglutamate aminopeptidase (PYR), leucine aminopeptidase (LAP), and esculin hydrolysis (ESC) activity
  • This test kit simplifies identification of Enterococcus spp. and group A streptococci (Streptococcus pyogenes)
  • Rapid:Only takes 10 minutes to complete
  • Easy and Simple:Easy to use format and simplifies the identification of Enterococcus spp. and Group A Streptococcu
  • Complete:Comes complete with cards, organism interpretive chart, and performs three relevant tests for gram-positive organisms
  • Convenient:All the reagents are ready-to-use and remain in one space saving workstation. Compact jar conserves precious refrigerator space
Specifications
Color Code Cherry red or bright pink is Positive for PYR and LAP, Light gray indicates positive reaction for ESC
Disposable Single-use
Analysis Time 10 min.
Form Liquid