Learn More
Bioworld Streptalon (Steptavidin Magnetic Beads) Kit, 2 mL
Catalog No. 50197618
Encompass_Preferred
bioWorld's Streptalon (Streptavidin magnetic beads) Kit is designed for convenient and quick purification of biotinylated proteins and nucleic acids inimmunoprecipitation, protein interaction studies, immunoassays and cell isolation, etc.Streptalon (Streptavidin Magnetic Beads)Kit providessuperparamagnetic beads of approximately 10 μm in size, covalently coated with highly purified streptavidin.Kit Contents:• Streptalon™ (25mg/ml magnetic beads), 2ml• Binding/Wash Buffer • Elution Buffer• Neutralization BufferThis kit requires MagneZoom™ paramagnetic bead station (20160000-1)for separation of beads.
Catalog No. 50-197-618 Supplier Bioworld Supplier No. 201630141
bioWorld's Streptalon (Streptavidin magnetic beads) Kit is designed for convenient and quick purification of biotinylated proteins and nucleic acids inimmunoprecipitation, protein interaction studies, immunoassays and cell isolation, etc.Streptalon (Streptavidin Magnetic Beads)Kit providessuperparamagnetic beads of approximately 10 μm in size, covalently coated with highly purified streptavidin.Kit Contents:• Streptalon™ (25mg/ml magnetic beads), 2ml• Binding/Wash Buffer • Elution Buffer• Neutralization BufferThis kit requires MagneZoom™ paramagnetic bead station (20160000-1)for separation of beads.