Learn More
Bioworld Streptalon (Streptavidin Magnetic Beads), 2 mL
Catalog No. 50197599
Encompass_Preferred
Streptavidin covalently linked to MagneZoom™ (magnetic beads). For magnetic separation and isolation of biotinylated ligands for affinity separations. Used in high throughput screening. Technical SpecificationsSurface silica coated magnetic beads Bead Size: 3-5 µm Supplied as suspension (25mg/ml)in phosphate buffer containing 0.1% BSA and 0.02 % sodium azide. Use MagneZoom paramagneic bead station (cat# 20160000-1) to capture beads.
Catalog No. 50-197-599 Supplier Bioworld Supplier No. 201620182
Streptavidin covalently linked to MagneZoom™ (magnetic beads). For magnetic separation and isolation of biotinylated ligands for affinity separations. Used in high throughput screening. Technical SpecificationsSurface silica coated magnetic beads Bead Size: 3-5 µm Supplied as suspension (25mg/ml)in phosphate buffer containing 0.1% BSA and 0.02 % sodium azide. Use MagneZoom paramagneic bead station (cat# 20160000-1) to capture beads.