Learn More
Medchemexpress LLC HY-P3152 1mg , Streptavidin CAS:9013-20-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP31521MG
Encompass_Preferred
HY-P3152 1mg Streptavidin CAS: 9013-20-1 Streptavidin is a ∼60 kDa homotetramer. Streptavidin binds four molecules of biotin with the highest affinity. The binding affinity of biotin to streptavidin is one of the highest reported for a non-covalent interaction to date, with a KD ∼ 0.01 pM. Streptavidin has an immunosuppressive role. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1993