Learn More
ACROBiosystems Streptavidin Coated Plates, Clear, 96-Well, Clear Frame (For Serological Testing) 1 plate
Supplier:  ACROBiosystems SP151PLATE
Encompass_Preferred
The plate could not be separated, and the frame is clear. Based on the high binding affinity between biotin-streptavidin systems, our best seller SP-15 Streptavidin Coated Plates (For Serological Testing) are recommended to use together with biotinylated proteins to get lower non-specific binding results in a shorter time and be easy to watch color changes.
Catalog No. 50-225-3651