Learn More
Rockland Immunochemicals STREPTAVIDIN - S000-01
SDP
Supplier:  Rockland Immunochemicals S00001
Encompass
Streptavidin is a bacterial protein (from Streptomyces avidinii) that has an exceptionally high binding affinity for biotin (B7). Streptavidin-biotin binding is one of the strongest known non-covalent interactions and is highly resistant to many conditions that would typically cause dissociation (such as organic solvents, denaturants, detergents, and extreme temperatures or pH). Streptavidin's affinity for biotin can be employed in a variety of experimental uses, from purifications to standards, to means of detection or pull down experiments.
Catalog No. 50-105-9142