Learn More
AdipoGen Streptavidin
SDP
Supplier:  AdipoGen AG35B0014C050
Encompass_Preferred
Recombinant protein. Recombinant streptavidin from Streptomyces avidinii. Binds to any biotinylated reagents. Endotoxin . Lyophilized. Contains PBS. Streptavidin is a non-glycosylated protein isolated from Streptomyces avidinii. It forms a homotetramer and strong non-covalent specific complex with biotin one molecule of biotin per subunit of streptavidin.
Catalog No. 50-256-3078