Learn More
ACROBiosystems Streptavidin (A24C) Protein, His Tag
Catalog No. 502718848
Encompass_Preferred
Streptavidin is a 66KDa tetrameric protein purified from the bacterium Streptomyces avidinii, and exhibits high binding affinity to biotin. Each unit can bind one biotin. Horseradish peroxidase is metalloenzyme, a 44KDa glycoprotein. When incubate with substrates, it produces a coloured, fluorimetric, or luminescent derivatives, which can be detected and quantified. HRP conjugated Streptavidin is widely used for the detection and quantification of biotinylated proteins.
Catalog No. 50-271-8848 Supplier ACROBiosystems Supplier No. STNN51H35MG
Streptavidin is a 66KDa tetrameric protein purified from the bacterium Streptomyces avidinii, and exhibits high binding affinity to biotin. Each unit can bind one biotin. Horseradish peroxidase is metalloenzyme, a 44KDa glycoprotein. When incubate with substrates, it produces a coloured, fluorimetric, or luminescent derivatives, which can be detected and quantified. HRP conjugated Streptavidin is widely used for the detection and quantification of biotinylated proteins.