Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptavidin Agarose, sedimented bead suspension

Catalog No. S951
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S951 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S951 Supplier Invitrogen™ Supplier No. S951
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The streptavidin conjugated to 4% beaded crosslinked agarose can be used to isolate biotinylated proteins.

The streptavidin conjugated to 4% beaded crosslinked agarose can be used to isolate biotinylated proteins. Additionally, biotinylated antibodies can be bound to streptavidin agarose to generate affinity matrices for the large-scale isolation of antigens.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Wet Ice
Form Liquid Suspension
Quantity 5 mL
Type Agarose
Label or Dye Agarose
Stationary Phase Streptavidin
Column Type Affinity
What is the binding capacity of streptavidin agarose?

The binding capacity of streptavidin agarose is 15-20 µg (18-24 nmoles) of fluorescein biotin per mL of sedimented gel.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.