Learn More
Vector Laboratories Protein ̶ Oligo Conjugation N-1000-002
SDP
Supplier:  Vector Laboratories N1000002
Encompass
Protein-Oligo Conjugation Kit. This kit is designed to easily and efficiently prepare two separate protein–oligo conjugates. This kit is flexible so that researchers with little or no conjugation experience can make their own custom protein–oligo conjugates to suit their research needs. It includes all of the necessary components, including S-HyNic and S-4FB, for the rapid and specific crosslinking of any protein with any oligo from 20 to 120 nucleotides in length. Up to 650 µg of protein may be conjugated to oligo in each reaction.
Catalog No. 50-204-5735