Learn More
Abcam Streptavidin AZDye™ 488 Monovalent Antibody Labeling Kit, 50UG
Catalog No. NC2064520
Encompass_Preferred
AZdye 488 conjugation / labeling in < 4 hrs with 30 secs hands-on time using Streptavidin AZDye™ 488 Monovalent Antibody Labeling Kit ab272187. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2064520 Supplier Abcam Supplier No. AB27218750UG
AZdye 488 conjugation / labeling in < 4 hrs with 30 secs hands-on time using Streptavidin AZDye™ 488 Monovalent Antibody Labeling Kit ab272187. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement