Learn More
Invitrogen™ Streptavidin, Alexa Fluor™ 635 conjugate
Description
View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
Specifications
Specifications
| Conjugate | Alexa Fluor 635 |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis |
| Quantity | 1 mg |
| Content And Storage | Contains 1 vial of streptavidin Alexa Fluor™ 635 conjugate. Store in freezer (-5 to -30°C) and protect from light. |
| Excitation/Emission | 632/647 |
| Flow Cytometer Laser Lines | 633 or 635 |
| Shipping Condition | Room Temperature |
| Form | Solid |
| Product Line | Alexa Fluor |
| Show More |
Frequently Asked Questions (FAQs)
In the lyophilized powder form, the fluorescent streptavidin labeled conjugate is stable for six months when stored at -20 degrees C, desiccated, and protected from light. The reconstituted solution is stable for approximately six months when stored at 4 degrees C, protected from light, with the addition of sodium azide to a final concentration of 5 mM or thimerosal to 0.2 mM. For longer storage, we recommend dividing the solution into aliquots and freezing at -20 degrees C, protected from light. Avoid repeated freezing and thawing of the solution.
The fluorescent streptavidin labeled conjugate solution can be made by dissolving the powder in 0.5-1.0 mL of PBS or other suitable buffer. For details, please refer to page 4 of the "Streptavidin and Fluorescent Conjugates of Streptavidin" manual (https://assets.fishersci.com/TFS-Assets/LSG/manuals/mp00888.pdf).
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.