Learn More
Abcam Streptavidin AZDye™ 647 Monovalent Antibody Labeling Kit, 50UG
Catalog No. NC2064519
Encompass_Preferred
AZdye 647 conjugation / labeling in < 4 hrs with 30 secs hands-on time using Streptavidin AZDye™ 647 Monovalent Antibody Labeling Kit ab272190. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2064519 Supplier Abcam Supplier No. AB27219050UG
AZdye 647 conjugation / labeling in < 4 hrs with 30 secs hands-on time using Streptavidin AZDye™ 647 Monovalent Antibody Labeling Kit ab272190. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement