Learn More
Sigma Aldrich Fine Chemicals Biosciences Streptavidin Alkaline Phos1MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S28901MG
Encompass
Streptavidin a 58 kDa protein is a homotetramer purified from Streptomyces avidinii. It has a high affinity for biotin. Streptavidin has been conjugated with either peroxidase or alkaline phosphatase (ALP). These enzymes are used as protein conjugates because of their comparative stability and a wide range of suitable substrates for different applications. ALP conjugates to streptavidinnbspin immunoassays.
Catalog No. 50-290-4576