Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptavidin, Alkaline Phosphatase Conjugate

Catalog No. S921
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S921 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S921 Supplier Invitrogen™ Supplier No. S921
Add to Cart
Edge
Add to Cart

Thermo Scientific Streptavidin, Alkaline Phosphatase Conjugate includes streptavidin in a purified form, conjugated to alkaline phosphatase for substrate based detection.

Thermo Scientific Streptavidin, Alkaline Phosphatase Conjugate includes streptavidin in a purified form, conjugated to alkaline phosphatase for substrate based detection.

Streptavidin is a biotin-binding protein originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Thermo Scientific streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.

Streptavidin, Alkaline Phosphatase Conjugate enables detection of biotinylated antibodies and other probes in a variety of standard assay methods, including western blotting, ELISA, immunohistochemistry, and fluorescence imaging.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 mg/mL
Conjugate AP
Quantity 0.5 mL
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Wet Ice
Form Liquid
Product Type Streptavidin Conjugate (fluorescent)
Label or Dye AP (Alkaline Phosphatase)
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.