Learn More
Agilent Technologies Streptavidin-Allophycocyanin (ver 2), 0.25 mg
Supplier:  Agilent Technologies PJ27S
Encompass
Streptavidin-Allophycocyanin (ver 2), 0.25 mg (formerly ProZyme). GT5 APC (Spirulina platensis) is crosslinked and conjugated to Streptavidin (Streptomyces avidinii). This product is specifically designed for use in tetramer and multimer applications.
Catalog No. NC1739572