Learn More
Invitrogen™ Streptavidin, Allophycocyanin Conjugate, premium grade
Description
View a selection guide for all streptavidin RPE, APC, and tandem conjugates.
Specifications
Specifications
| Concentration | 1 mg/mL |
| Conjugate | APC |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis |
| Quantity | 250 μL |
| Content And Storage | Contains 1 vial of streptavidin APC, premium grade, conjugate (1 mg/mL). Store in refrigerator (2–8°C) and protect from light. |
| Excitation/Emission | 650/660 |
| Flow Cytometer Laser Lines | 633 or 635 |
| Shipping Condition | Wet Ice |
| Form | Solution |
| Show More |
Frequently Asked Questions (FAQs)
Streptavidin APC, premium grade has a degree of labeling of one APC molecule to one streptavidin molecule.
Allophycocyanin-conjugated antibodies must be stored at 2-8 degrees C. Antibodies conjugated to phycobiliproteins (APC, R-PE) must never be frozen. Once frozen, even briefly, they are no longer usable.
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.