Learn More
Seracare Life Sciences Inc STREPTAVIDIN AP-LABELED
Supplier:  Seracare Life Sciences Inc 52700030
Encompass
Streptavidin labeled with alkaline phosphatase. Streptavidin is a protein isolated from the bacterium Streptomyces avidinii and has no carbohydrate moieties. It has a molecular weight of approximately 60,000 Daltons with an isoelectric point of 5. Product is in lyophilized form.
Catalog No. 50-672-53