Learn More
Columbia Biosciences Corp STREPTAVIDIN APC 1MG
Supplier:  Columbia Biosciences Corp D32212
Encompass_Preferred
Streptavidin conjugated to SureLight Allophycocyanin, 100ug Optimized for High throughput screening (HTS) and fluorescence resonance energy transfer (FRET) assays targeting kinase activity, flow cytometry
Catalog No. 50-921-428