Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptavidin, (APC-Alexa Fluor™ 750)

Catalog No. SA1027
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA1027 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA1027 Supplier Invitrogen™ Supplier No. SA1027
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Streptavidin, (APC-Alexa Fluor™ 750)

  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide

View a selection guide for all streptavidin RPE, APC, and tandem conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Conjugate APC-Alexa Fluor 750
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Quantity 1 mL
Packaging Tube
Content And Storage Contains 1 vial of streptavidin, APC-Alexa Fluor™ 750 conjugate.

Store in refrigerator (2°C to 8°C).
Excitation/Emission 600-650/775 nm
Flow Cytometer Laser Lines 633 or 635
Shipping Condition Wet Ice
Form Solution
Product Line Alexa Fluor
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye APC-Alexa Fluor 750
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.