Learn More
Description
- For flow cytometry, fluorescence microscopy and imaging.
- Exhibits significantly improved brightness.
Specifications
Specifications
| Antigen | Streptavidin |
| Concentration | 0.1 mg/mL |
| Conjugate | APC-R700 |
| Quantity | 0.1 mg |
| Regulatory Status | For research use only (RUO) |
| Storage Buffer | Aqueous buffered solution containing protein stabilizer and less than or equal to 0.09% sodium azide |
| Content And Storage | Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze. |
| For Use With (Application) | Flow Cytometry |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.