Learn More
ACROBiosystems Streptavidin Beads 100mg
Catalog No. 502534461
Encompass_Preferred
ActiveMax Streptavidin coupled-ubeads premium grade (for cells) are produced under sterile manufacturing conditions (ISO 5) and no animal- or human-derived components are used throughout the production process. It is produced under our rigorous quality control system that includes a comprehensive set of tests including sterility and endotoxin tests.
Catalog No. 50-253-4461 Supplier ACROBiosystems Supplier No. MBSC009100MG
ActiveMax Streptavidin coupled-ubeads premium grade (for cells) are produced under sterile manufacturing conditions (ISO 5) and no animal- or human-derived components are used throughout the production process. It is produced under our rigorous quality control system that includes a comprehensive set of tests including sterility and endotoxin tests.