Learn More
ACROBiosystems Magnetic Beads, Streptavidin, 2.8 μm, Lyophilized Powder, 10 mg
Catalog No. 502331086
Encompass_Preferred
These 2.8 μm superparamagnetic particles are covalently coupled with highly pure streptavidin for the efficient capture of biotinylated proteins and molecules. Featuring an extremely high binding affinity for biotin (Kd ~10-14 mol/L), these beads facilitate irreversible binding while minimizing non-specific interactions due to their optimized isoelectric point. Designed for chemiluminescence procedures, they provide a ready-to-use solution that preserves protein activity and ensures reproducible results.
  • Particle size: 2.8 μm
  • Surface chemistry: Hydrophilic
  • Binding capacity: 5-20 μg biotinylated protein per mg of beads
  • Lower non-specific binding compared to avidin
  • Optimized for chemiluminescence applications
Catalog No. 50-233-1086 Supplier ACROBiosystems Supplier No. MPCA00610MG
These 2.8 μm superparamagnetic particles are covalently coupled with highly pure streptavidin for the efficient capture of biotinylated proteins and molecules. Featuring an extremely high binding affinity for biotin (Kd ~10-14 mol/L), these beads facilitate irreversible binding while minimizing non-specific interactions due to their optimized isoelectric point. Designed for chemiluminescence procedures, they provide a ready-to-use solution that preserves protein activity and ensures reproducible results.
  • Particle size: 2.8 μm
  • Surface chemistry: Hydrophilic
  • Binding capacity: 5-20 μg biotinylated protein per mg of beads
  • Lower non-specific binding compared to avidin
  • Optimized for chemiluminescence applications