Learn More
Apexbio Technology LLC Streptavidin Beads T3,1ml
SDP
Catalog No. 502239983
Encompass_Preferred
APExBIO Streptavidin Beads T3 are ideal for numerous applications, including purification of proteins and nucleic acids, protein interaction studies, immunoprecipitation, immunoassays, phage display, biopanning, drug screening and cell isolation.Add the beads to a sample containing biotinylated molecules, e.g. peptides, proteins, antibodies, sugars, lectins, oligonucleotides, DNA/RNA. During a short incubation, the biotinylated molecule will bind to the beads. The molecule-bead complex can be separated by using a magnet. capture, washing, and detection can be optimized for manual or automated use. With indirect capture, mix the biotinylated molecule with the sample to capture the molecule-target complex before adding the beads. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-223-9983 Supplier Apexbio Technology LLC Supplier No. K13011
APExBIO Streptavidin Beads T3 are ideal for numerous applications, including purification of proteins and nucleic acids, protein interaction studies, immunoprecipitation, immunoassays, phage display, biopanning, drug screening and cell isolation.Add the beads to a sample containing biotinylated molecules, e.g. peptides, proteins, antibodies, sugars, lectins, oligonucleotides, DNA/RNA. During a short incubation, the biotinylated molecule will bind to the beads. The molecule-bead complex can be separated by using a magnet. capture, washing, and detection can be optimized for manual or automated use. With indirect capture, mix the biotinylated molecule with the sample to capture the molecule-target complex before adding the beads. Other sizes are also available. Please inqury us for quote.