Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Streptavidin, BV421

Catalog No. BDB563259 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563259 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563259 Supplier BD Supplier No. 563259
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

100μg in 0.1mg/mL concentration

Species Reactivity:Ms

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.1 mg/mL
Quantity 100 μg
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.