Learn More
XpressBio Xpress Capture Streptavidin Coated ELISA Plates, 96 White Wells (5-pack)
SDP
Supplier:  XpressBio EX8000W96
Encompass_Preferred
Capture coated Streptavidin ELISA plates may be used with biotinylated peptides, antigens, antibodies, DNA, and oligonucleotides for various uses such as immunoassays and DNA/RNA quantitation.
Catalog No. 50-171-7654