Learn More
Bioworld Streptavidin-Coated Microplates, 5 Plate(s)
Catalog No. 50197542
Encompass
Streptavidin Coated Plates are available in 96-wellplate format, and pre-blocked with BSA Blocking Buffer.bio-world Biotin-Binding Plates are available in clear to be used with colorimetricdetection methods. Specifications:Easy and gentle immobilization of biotin-containing conjugates Low nonspecific binding No denaturing of the protein component of a conjugate upon binding Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene Pre-blocked with BSA Buffer
Catalog No. 50-197-542 Supplier Bioworld Supplier No. 201400111
Streptavidin Coated Plates are available in 96-wellplate format, and pre-blocked with BSA Blocking Buffer.bio-world Biotin-Binding Plates are available in clear to be used with colorimetricdetection methods. Specifications:Easy and gentle immobilization of biotin-containing conjugates Low nonspecific binding No denaturing of the protein component of a conjugate upon binding Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene Pre-blocked with BSA Buffer