Learn More
Spherotech STREPTAVIDIN COATED POLYSTYREN
Catalog No. NC0823438
Encompass_Preferred
Streptavidin Polystyrene Particles, 0.5% w/v, 4.0-4.9 um, 5 mL
Catalog No. NC0823438 Supplier Spherotech Supplier No. SVP405
Streptavidin Polystyrene Particles, 0.5% w/v, 4.0-4.9 um, 5 mL