Learn More
Spherotech STREPTAVIDIN COATED PS PARTICL
Catalog No. NC0320924
Encompass_Preferred
Streptavidin Polystyrene Particles, 0.5% w/v, 6.0-8.0 um, 5 mL
Catalog No. NC0320924 Supplier Spherotech Supplier No. SVP605
Streptavidin Polystyrene Particles, 0.5% w/v, 6.0-8.0 um, 5 mL