Learn More
Abcam Streptavidin Conjugation Kit - Lightning-Link, 3 x 10UG
Catalog No. 502585575
Encompass_Preferred
The Abcam Streptavidin Conjugation Kit (ab102921) offers several standout features: -Rapid Conjugation: Achieve streptavidin labeling in under 4 hours with just 30 seconds of hands-on time. -High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. -Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification. - Confidence: cited in over 20 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5575 Supplier Abcam Supplier No. AB1029213X10UG
The Abcam Streptavidin Conjugation Kit (ab102921) offers several standout features: -Rapid Conjugation: Achieve streptavidin labeling in under 4 hours with just 30 seconds of hands-on time. -High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. -Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification. - Confidence: cited in over 20 publications
The product is subject to the following: Abcam Restricted Use Statement