Learn More
Spherotech STREPTAVIDIN CT NILE RED PARTI
Catalog No. NC0516145
Encompass_Preferred
Streptavidin Fluorescent Particles, Nile Red, 0.1% w/v, 5.0-7.9 um, 5 mL
Catalog No. NC0516145 Supplier Spherotech Supplier No. SVFP60565
Streptavidin Fluorescent Particles, Nile Red, 0.1% w/v, 5.0-7.9 um, 5 mL