Learn More
Apexbio Technology LLC Streptavidin - Cy3, 2x1mL(0.5mg/mL). Cas: MFCD: NA. Used in fluorescent detection of biotinylated antibodies, proteins or other molecules
SDP
Catalog No. 501998212
Encompass_Preferred
Streptavidin-Cy3 is useful for the detection of biotinylated antibodies, proteins, nucleic acids or other molecules in immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), in situ hybridization (ISH), or flow cytometry. Concentration: 0.5mg/mL. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8212 Supplier Apexbio Technology LLC Supplier No. K10792
Streptavidin-Cy3 is useful for the detection of biotinylated antibodies, proteins, nucleic acids or other molecules in immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), in situ hybridization (ISH), or flow cytometry. Concentration: 0.5mg/mL. Other sizes are also available. Please inqury us for quote.