Learn More
Vector Laboratories Alkaline Phosphatase Avidin D SA-1300-1
SDP
Supplier:  Vector Laboratories SA13001
Encompass
Alkaline Phosphatase Avidin D, 100U, is a powerful tool to detect or purify biotinylated proteins, nucleic acids, and other macromolecules. Vector Laboratories enzyme-conjugated avidin and streptavidin are produced with the highest specific activity enzymes in optimal ratios. Specific covalent linkages are chosen to provide stable, highly active conjugates. These enzyme conjugates are suitable for use in solid-phase assays, tissue/cell staining systems, and blotting applications. One unit is 1 mole p-nitrophenylphosphate hydrolyzed per minute at 37 degrees C, pH 9.8.
Catalog No. 50-204-5699