Learn More
Sigma Aldrich Fine Chemicals Biosciences Streptavidin Cy3TM from St
Supplier: Sigma Aldrich Fine Chemicals Biosciences S64021ML
Streptavidin is a homotetramer protein. One molecule of streptavidin binds four molecules of biotin by a non-covalent interaction that is essentially irreversible.