Learn More
Seracare Life Sciences Inc STREPTAVIDIN CY5-LABELED
Supplier:  Seracare Life Sciences Inc 52700023
Encompass_Preferred
Streptavidin is a 60,000 Dalton protein isolated from the bacterium Streptomyces avidinii. Electrophoretically pure streptavidin was labeled with Cy5 using a proprietary method. DyLight conjugates provide increased brightness and greater protostability than other fluorophores (ex. - FITC). The conjugate is designed for use in immunofluorescense assays including flow cytometry, immunohistology, Western blotting, microarrays and FLISA.
Catalog No. 50-670-73