Learn More
Seracare Life Sciences Inc STREPTAVIDIN DYLIGHT 488
Supplier:  Seracare Life Sciences Inc 52700017
Encompass_Preferred
Streptavidin is a 60,000 Dalton protein isolated from the bacterium Streptomyces avidinii. Electrophoretically pure streptavidin was labeled with DyLight 488 using a proprietary method. DyLight conjugates provide increased brightness.
Catalog No. 50-669-52