Learn More
Invitrogen™ Streptavidin, β-galactosidase conjugate
Description
The streptavidin, ß-galactosidase conjugate can be used to detect biotin in a signal amplification scheme in conjunction with chromogenic substrates or fluorogenic substrates.
Specifications
Specifications
| Conjugate | beta-Galactosidase |
| Quantity | 1 mg |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Shipping Condition | Wet Ice |
| Form | Solid |
| Product Type | Streptavidin Conjugate (fluorescent) |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.