Learn More
Apexbio Technology LLC Streptavidin-HyperFluor 555 1mL(0.5mg/mL)
SDP
Catalog No. 503107186
Encompass_Preferred
Streptavidin can be used for the detection of biotin-labeled antibodies nucleic acids proteins or other biotinylated molecules HyperFluor 555 is a commonly used fluorescent dye widely applied in biological and medical research particularly in techniques such as fluorescence microscopy flow cytometry and fluorescence resonance energy transfer (FRET) It exhibits greenish-yellow fluorescence with excitation and emission wavelengths around 555 nm and 565 nm respectively Streptavidin-HyperFluor 555 is a bioprobe conjugate comprising streptavidin protein and the HyperFluor 555 fluorescent dye serving as a powerful and sensitive fluorescent labeling tool
Catalog No. 50-310-7186 Supplier Apexbio Technology LLC Supplier No. K44041
Add to Cart
Add to Cart
Streptavidin can be used for the detection of biotin-labeled antibodies nucleic acids proteins or other biotinylated molecules HyperFluor 555 is a commonly used fluorescent dye widely applied in biological and medical research particularly in techniques such as fluorescence microscopy flow cytometry and fluorescence resonance energy transfer (FRET) It exhibits greenish-yellow fluorescence with excitation and emission wavelengths around 555 nm and 565 nm respectively Streptavidin-HyperFluor 555 is a bioprobe conjugate comprising streptavidin protein and the HyperFluor 555 fluorescent dye serving as a powerful and sensitive fluorescent labeling tool