Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptavidin, horseradish peroxidase conjugate

Catalog No. S911A
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S911A 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S911A Supplier Invitrogen™ Supplier No. S911
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The streptavidin, horseradish peroxidase (HRP) conjugate can be used to detect biotin in a signal amplification scheme in conjunction with chromogenic substrates or fluorogenic HRP substrates.

The streptavidin, horseradish peroxidase (HRP) conjugate can be used to detect biotin in a signal amplification scheme in conjunction with chromogenic substrates or fluorogenic HRP substrates, such as the tyramide signal amplification (TSA) technology.

We recommend reconstituting in 1 mL of 1X phosphate-buffered saline (PBS), pH 7.2, for a 1 mg/mL stock solution.

TRUSTED_SUSTAINABILITY

Specifications

Conjugate HRP
Quantity 1 mg
Content And Storage Store in freezer (-5°C to -30°C).
Shipping Condition Wet Ice
Product Type Streptavidin Conjugate (fluorescent)
Label or Dye HRP
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.