Learn More
Abcam Streptavidin (HRP), 500UG
Supplier:  Abcam AB7403500UG
Encompass_Preferred
Streptavidin (HRP) is a Full Length native Streptavidin protein expressed in Streptomyces avidinii and conjuated with Horseradish peroxidase (HRP). ab7403 has >95% purity and suitable for western blot, ELISA, IHC, ICC, Dot blot and more. - Used to detect biotin-conjugated primary antibodies - Has less non-specific binding and less background signal compared to avidin - Cited in over 170 publications
Protein/ Peptide/ Enzyme Synonyms: Streptavidin
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC9689400