Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Streptavidin HRP Solution

Catalog No. bdb554066 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554066 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554066 Supplier BD Supplier No. 554066
Only null left
Add to Cart
Add to Cart

Streptavidin is a non-glycosylated protein that is prepared chromatographically from the bacterium Streptomyces avidinii. Streptavidin homotetramers have a particularly high, non-covalent binding affinity for biotin.

  • Each investigator should titrate the reagent to obtain optimal results
  • Use in ELISA, immunocytochemistry, and immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Conjugate HRP
pH 7.2
Quantity 1 mL
Content And Storage Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze.
Form Liquid
For Use With (Application) ELISA
Buffer Aqueous buffered solution containing BSA and ProClin™ 150 as preservative
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.